CAS RN 7237-34-5
IUPAC Name:
(2-hydroxyethyl)triphenylphosphanium bromide
Synonyms:
(2-hydroxyethyl)triphenylphosphanium bromide
Molecular Weight (g/mol):
387.26
Molecular Formula:
C20H20BrOP
InChi Key:
QZJOQNHOOVSESC-UHFFFAOYSA-M
SMILES:
[Br-].OCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
Promotions Available
(2-Hydroxyethyl)triphenylphosphonium bromide, 98+%
(2-Hydroxyethyl)triphenylphosphonium bromide, 98+%
CAS: 7237-34-5 Molecular Formula: C20H20BrOP Molecular Weight (g/mol): 387.26 MDL Number: MFCD00011901 InChI Key: QZJOQNHOOVSESC-UHFFFAOYSA-M Synonym: 2-hydroxyethyl triphenylphosphonium bromide,2-hydroxyethyl triphenylphosphanium bromide,acmc-20appe,2-hydroxyethyltriphenylphosphonium bromide,2-hydroxyethyl triphenyphosphonium bromide,2-hydroxyethyl triphenyl phosphanium bromide,2-hydroxy ethyl triphenylphosphonium bromide,phosphonium, 2-hydroxyethyl triphenyl-,bromide 1:1,phosphonium, 2-hydroxyethyl triphenyl-, bromide 1:1 PubChem CID: 2733550 IUPAC Name: 2-hydroxyethyl(triphenyl)phosphanium;bromide SMILES: [Br-].OCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1