missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxybenzophenone 95.0+%, TCI America™

Catalog No. H04705G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H04705G 5 g
H047025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H04705G Supplier TCI America Supplier No. H04705G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 117-99-7
Molecular Formula C13H10O2
Molecular Weight (g/mol) 198.221
MDL Number MFCD00002216
InChI Key HJIAMFHSAAEUKR-UHFFFAOYSA-N
Synonym 2-hydroxybenzophenone, o-benzoylphenol, 2-hydroxyphenyl phenyl methanone, o-hydroxybenzophenone, benzophenone, 2-hydroxy, methanone, 2-hydroxyphenyl phenyl, phenyl 2-hydroxyphenyl ketone, unii-lqv22w4v05, ortho-hydroxybenzophenone, 2-hydroxyphenyl phenylmethanone
PubChem CID 8348
IUPAC Name (2-hydroxyphenyl)-phenylmethanone
SMILES C1=CC=C(C=C1)C(=O)C2=CC=CC=C2O

Specifications

Melting Point 37°C
Boiling Point 148°C
Quantity 5 g
Formula Weight 198.22
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxybenzophenone
TSCA Yes
Recommended Storage Refrigerator