accessibility menu, dialog, popup

UserName

OHO

CAS RN 117-99-7

IUPAC Name: 2-benzoylphenol
Synonyms: O-hydroxybenzophenone
Molecular Weight (g/mol): 198.22
Molecular Formula: C13H10O2
InChi Key: HJIAMFHSAAEUKR-UHFFFAOYSA-N
SMILES: OC1=CC=CC=C1C(=O)C1=CC=CC=C1
Boiling Point 
  • (2)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (4)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (3)
  • (2)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (3)
  • (4)
  • (2)
  • (5)
  • (1)
  • (1)
  • (3)
  • (2)
  • (5)