missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxybenzoic Acid 99.5+%, TCI America™

Supplier:  TCI America H13425G

 View more versions of this product

Catalog No. H13425G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-Hydroxybenzoic Acid [for Biochemical Research]
159°C
211°C
MFCD00002439
salicylic acid, o-hydroxybenzoic acid, 2-carboxyphenol, o-carboxyphenol, rutranex, salonil, retarder w, keralyt, duoplant, freezone
OC(=O)C1=CC=CC=C1O
138.12
CHEBI:16914
≥99.5% (T)
69-72-7
White
C7H6O3
5 g
YGSDEFSMJLZEOE-UHFFFAOYSA-N
2-hydroxybenzoic acid
338
138.12
Crystalline Powder

Chemical Identifiers

69-72-7
138.12
YGSDEFSMJLZEOE-UHFFFAOYSA-N
338
2-hydroxybenzoic acid
C7H6O3
MFCD00002439
salicylic acid, o-hydroxybenzoic acid, 2-carboxyphenol, o-carboxyphenol, rutranex, salonil, retarder w, keralyt, duoplant, freezone
CHEBI:16914
OC(=O)C1=CC=CC=C1O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1640

RTECSNumber : VO0525000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions