missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxybenzoic Acid 99.5+%, TCI America™

Catalog No. p-7130993
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H13425G 5 g
H134225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H13425G Supplier TCI America Supplier No. H13425G
Only null left

Chemical Identifiers

CAS 69-72-7
Molecular Formula C7H6O3
Molecular Weight (g/mol) 138.12
MDL Number MFCD00002439
InChI Key YGSDEFSMJLZEOE-UHFFFAOYSA-N
Synonym salicylic acid, o-hydroxybenzoic acid, 2-carboxyphenol, o-carboxyphenol, rutranex, salonil, retarder w, keralyt, duoplant, freezone
PubChem CID 338
ChEBI CHEBI:16914
IUPAC Name 2-hydroxybenzoic acid
SMILES OC(=O)C1=CC=CC=C1O

Specifications

Melting Point 159°C
Color White
Boiling Point 211°C
Quantity 5 g
Formula Weight 138.12
Percent Purity ≥99.5% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxybenzoic Acid [for Biochemical Research]
EINECSNumber (3)-1640
RTECSNumber VO0525000
TSCA Yes