missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Hydroxybenzoic Acid 99.5+%, TCI America™
Supplier: TCI America H134225G
Specifications
| 2-Hydroxybenzoic Acid [for Biochemical Research] | |
| 159°C | |
| 211°C | |
| MFCD00002439 | |
| salicylic acid, o-hydroxybenzoic acid, 2-carboxyphenol, o-carboxyphenol, rutranex, salonil, retarder w, keralyt, duoplant, freezone | |
| OC(=O)C1=CC=CC=C1O | |
| 138.12 | |
| CHEBI:16914 | |
| ≥99.5% (T) |
| 69-72-7 | |
| White | |
| C7H6O3 | |
| 25 g | |
| YGSDEFSMJLZEOE-UHFFFAOYSA-N | |
| 2-hydroxybenzoic acid | |
| 338 | |
| 138.12 | |
| Crystalline Powder |
Chemical Identifiers
| 69-72-7 | |
| 138.12 | |
| YGSDEFSMJLZEOE-UHFFFAOYSA-N | |
| 338 | |
| 2-hydroxybenzoic acid |
| C7H6O3 | |
| MFCD00002439 | |
| salicylic acid, o-hydroxybenzoic acid, 2-carboxyphenol, o-carboxyphenol, rutranex, salonil, retarder w, keralyt, duoplant, freezone | |
| CHEBI:16914 | |
| OC(=O)C1=CC=CC=C1O |
Safety and Handling
EINECSNumber : (3)-1640
RTECSNumber : VO0525000
TSCA : Yes