missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-5-nitrobenzyl Alcohol 98.0+%, TCI America™

Catalog No. H15495G
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H15495G 5 g
H154925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. H15495G missing translation for 'mfr' TCI America missing translation for 'supplierNo' H15495G
Only null left

Chemical Identifiers

CAS 39224-61-8
Molecular Formula C7H7NO4
Molecular Weight (g/mol) 169.14
MDL Number MFCD01321361
InChI Key JEIYIXDNZKATLC-UHFFFAOYSA-N
Synonym 5-Nitrosalicyl Alcohol
PubChem CID 2763823
IUPAC Name 2-(hydroxymethyl)-4-nitrophenol
SMILES OCC1=CC(=CC=C1O)[N+]([O-])=O

Specifications

Melting Point 125°C
Color Yellow
Quantity 5 g
Formula Weight 169.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxy-5-nitrobenzyl Alcohol