accessibility menu, dialog, popup

UserName

HOHONOO

CAS RN 39224-61-8

IUPAC Name: 2-(hydroxymethyl)-4-nitrophenol
Synonyms: 2-(hydroxymethyl)-4-nitrophenol
Molecular Weight (g/mol): 169.14
Molecular Formula: C7H7NO4
InChi Key: JEIYIXDNZKATLC-UHFFFAOYSA-N
SMILES: OCC1=CC(=CC=C1O)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results