missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-5-nitro-m-anisaldehyde 99.0+%, TCI America™

Catalog No. H03975G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H03975G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H03975G Supplier TCI America Supplier No. H03975G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 17028-61-4
Molecular Formula C8H7NO5
Molecular Weight (g/mol) 197.15
MDL Number MFCD00017033
InChI Key HGKHVFKBOHFYSS-UHFFFAOYSA-N
Synonym 3-methoxy-5-nitrosalicylaldehyde, 2-hydroxy-5-nitro-m-anisaldehyde, benzaldehyde, 2-hydroxy-3-methoxy-5-nitro, 5-nitro-o-vanillin, acmc-1bt2m, 3-methoxy-5-nitrosalicyladehyde, 2-hydroxy-5-nitro-3-methoxybenzaldehyde, benzaldehyde,2-hydroxy-3-methoxy-5-nitro
PubChem CID 307886
IUPAC Name 2-hydroxy-3-methoxy-5-nitrobenzaldehyde
SMILES COC1=CC(=CC(C=O)=C1O)[N+]([O-])=O

Specifications

Melting Point 140°C
Color Yellow
Quantity 5 g
Formula Weight 197.15
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxy-5-nitro-m-anisaldehyde