accessibility menu, dialog, popup

UserName

CAS RN 17028-61-4

IUPAC Name: 2-hydroxy-3-methoxy-5-nitrobenzaldehyde
Synonyms: 2-hydroxy-3-methoxy-5-nitrobenzaldehyde
Molecular Weight (g/mol): 197.15
Molecular Formula: C8H7NO5
InChi Key: HGKHVFKBOHFYSS-UHFFFAOYSA-N
SMILES: COC1=CC(=CC(C=O)=C1O)[N+]([O-])=O
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)