missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2'-Hydroxy-5'-methylacetanilide 98.0+%, TCI America™
$52.19 - $130.48
Chemical Identifiers
| CAS | 6375-17-3 |
|---|---|
| Molecular Formula | C9H11NO2 |
| Molecular Weight (g/mol) | 165.19 |
| MDL Number | MFCD00020183 |
| InChI Key | DSEQJUPGRWESKP-UHFFFAOYSA-N |
| Synonym | 2-acetamido-4-methylphenol, 2-hydroxy-5-methylacetanilide, n-2-hydroxy-5-methylphenyl acetamide, 2'-hydroxy-5'-methylacetanilide, acetamide, n-2-hydroxy-5-methylphenyl, 2-acetamido-4-cresol, n1-2-hydroxy-5-methylphenyl acetamide, 2-acetamido-p-cresol, 6'-hydroxy-m-acetotoluidide, n-2-hydroxy-5-methyl-phenyl acetamide |
| PubChem CID | 292583 |
| IUPAC Name | N-(2-hydroxy-5-methylphenyl)acetamide |
| SMILES | CC(=O)NC1=CC(C)=CC=C1O |
Chemical Identifiers
| 6375-17-3 | |
| 165.19 | |
| DSEQJUPGRWESKP-UHFFFAOYSA-N | |
| 292583 | |
| CC(=O)NC1=CC(C)=CC=C1O |
| C9H11NO2 | |
| MFCD00020183 | |
| 2-acetamido-4-methylphenol, 2-hydroxy-5-methylacetanilide, n-2-hydroxy-5-methylphenyl acetamide, 2'-hydroxy-5'-methylacetanilide, acetamide, n-2-hydroxy-5-methylphenyl, 2-acetamido-4-cresol, n1-2-hydroxy-5-methylphenyl acetamide, 2-acetamido-p-cresol, 6'-hydroxy-m-acetotoluidide, n-2-hydroxy-5-methyl-phenyl acetamide | |
| N-(2-hydroxy-5-methylphenyl)acetamide |
Specifications
| 6375-17-3 | |
| Yellow | |
| MFCD00020183 | |
| 2-acetamido-4-methylphenol, 2-hydroxy-5-methylacetanilide, n-2-hydroxy-5-methylphenyl acetamide, 2'-hydroxy-5'-methylacetanilide, acetamide, n-2-hydroxy-5-methylphenyl, 2-acetamido-4-cresol, n1-2-hydroxy-5-methylphenyl acetamide, 2-acetamido-p-cresol, 6'-hydroxy-m-acetotoluidide, n-2-hydroxy-5-methyl-phenyl acetamide | |
| CC(=O)NC1=CC(C)=CC=C1O | |
| 165.19 | |
| 165.19 | |
| Crystalline Powder |
| 160°C | |
| C9H11NO2 | |
| 5 g | |
| DSEQJUPGRWESKP-UHFFFAOYSA-N | |
| N-(2-hydroxy-5-methylphenyl)acetamide | |
| 292583 | |
| ≥98.0% (HPLC,N) | |
| 2′-Hydroxy-5′-methylacetanilide |
Safety and Handling
EINECSNumber : (3)-0708
TSCA : Yes