accessibility menu, dialog, popup

UserName

CAS RN 6375-17-3

CH3ONHCH3HO

CAS RN 6375-17-3

IUPAC Name: N-(2-hydroxy-5-methylphenyl)acetamide
Synonyms: N-(2-hydroxy-5-methylphenyl)acetamide
Molecular Weight (g/mol): 165.19
Molecular Formula: C9H11NO2
InChi Key: DSEQJUPGRWESKP-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC(C)=CC=C1O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results