missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-5-methoxybenzaldehyde, 98%

CAS: 672-13-9 | C8H8O3 | 152.149 g/mol

Supplier:  Thermo Scientific Chemicals A1575318

 View more versions of this product

Catalog No. AAA1575318


Only null left
Explore available promotions

Description

Description

2-Hydroxy-5-methoxybenzaldehyde is employed to study electroantennogram response of the vine weevil (Otiorhynchus sulcatus F) to a broad range of volatile plant compounds. It plays an important role in the preparation of tetradentate Schiff base compounds. Further, it is used in the preparation 6-methoxy-3-nitro-2H-chromene by using dibutylamine as a reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hydroxy-5-methoxybenzaldehyde is employed to study electroantennogram response of the vine weevil (Otiorhynchus sulcatus F) to a broad range of volatile plant compounds. It plays an important role in the preparation of tetradentate Schiff base compounds. Further, it is used in the preparation 6-methoxy-3-nitro-2H-chromene by using dibutylamine as a reagent.

Solubility
Miscible with chloroform. Slightly miscible with water.

Notes
Air sensitive. Incompatible with strong oxidizing agents and strong bases.
Specifications

Specifications

2-Hydroxy-5-methoxybenzaldehyde
13°C to 16°C
248°C to 252°C
C8H8O3
MFCD00003332
1100532
5-methoxysalicylaldehyde, m-anisaldehyde, 6-hydroxy, benzaldehyde, 2-hydroxy-5-methoxy, salicylaldehyde, 5-methoxy, 6-hydroxy-m-anisaldehyde, 2-hydroxy-5-methoxy-benzaldehyde, 5-methoxy-2-hydroxybenzaldehyde, 2-hydroxy-5-methoxy benzaldehyde, 2-formyl-4-methoxyphenol, pubchem2649
FZHSPPYCNDYIKD-UHFFFAOYSA-N
2-hydroxy-5-methoxybenzaldehyde
95695
98%
672-13-9
1.221
>110°C (230°F)
1.578
50 g
Air Sensitive
Miscible with chloroform. Slightly miscible with water.
COC1=CC(=C(C=C1)O)C=O
152.149
152.15

Chemical Identifiers

672-13-9
152.149
FZHSPPYCNDYIKD-UHFFFAOYSA-N
95695
COC1=CC(=C(C=C1)O)C=O
C8H8O3
MFCD00003332
5-methoxysalicylaldehyde, m-anisaldehyde, 6-hydroxy, benzaldehyde, 2-hydroxy-5-methoxy, salicylaldehyde, 5-methoxy, 6-hydroxy-m-anisaldehyde, 2-hydroxy-5-methoxy-benzaldehyde, 5-methoxy-2-hydroxybenzaldehyde, 2-hydroxy-5-methoxy benzaldehyde, 2-formyl-4-methoxyphenol, pubchem2649
2-hydroxy-5-methoxybenzaldehyde
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 211-589-0

RTECSNumber : BZ2800000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only