Learn More
2-Hydroxy-5-methoxybenzaldehyde, 98%
CAS: 672-13-9 | C8H8O3 | 152.149 g/mol
Supplier: Thermo Scientific Chemicals A1575314
Description
2-Hydroxy-5-methoxybenzaldehyde is employed to study electroantennogram response of the vine weevil (Otiorhynchus sulcatus F) to a broad range of volatile plant compounds. It plays an important role in the preparation of tetradentate Schiff base compounds. Further, it is used in the preparation 6-methoxy-3-nitro-2H-chromene by using dibutylamine as a reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Hydroxy-5-methoxybenzaldehyde is employed to study electroantennogram response of the vine weevil (Otiorhynchus sulcatus F) to a broad range of volatile plant compounds. It plays an important role in the preparation of tetradentate Schiff base compounds. Further, it is used in the preparation 6-methoxy-3-nitro-2H-chromene by using dibutylamine as a reagent.
Solubility
Miscible with chloroform. Slightly miscible with water.
Notes
Air sensitive. Incompatible with strong oxidizing agents and strong bases.
Specifications
| 2-Hydroxy-5-methoxybenzaldehyde | |
| 13°C to 16°C | |
| 248°C to 252°C | |
| C8H8O3 | |
| MFCD00003332 | |
| 1100532 | |
| 5-methoxysalicylaldehyde, m-anisaldehyde, 6-hydroxy, benzaldehyde, 2-hydroxy-5-methoxy, salicylaldehyde, 5-methoxy, 6-hydroxy-m-anisaldehyde, 2-hydroxy-5-methoxy-benzaldehyde, 5-methoxy-2-hydroxybenzaldehyde, 2-hydroxy-5-methoxy benzaldehyde, 2-formyl-4-methoxyphenol, pubchem2649 | |
| FZHSPPYCNDYIKD-UHFFFAOYSA-N | |
| 2-hydroxy-5-methoxybenzaldehyde | |
| 95695 | |
| 98% |
| 672-13-9 | |
| 1.221 | |
| >110°C (230°F) | |
| 1.578 | |
| 25 g | |
| Air Sensitive | |
| Miscible with chloroform. Slightly miscible with water. | |
| COC1=CC(=C(C=C1)O)C=O | |
| 152.149 | |
| 152.15 |
Chemical Identifiers
| 672-13-9 | |
| 152.149 | |
| FZHSPPYCNDYIKD-UHFFFAOYSA-N | |
| 95695 | |
| COC1=CC(=C(C=C1)O)C=O |
| C8H8O3 | |
| MFCD00003332 | |
| 5-methoxysalicylaldehyde, m-anisaldehyde, 6-hydroxy, benzaldehyde, 2-hydroxy-5-methoxy, salicylaldehyde, 5-methoxy, 6-hydroxy-m-anisaldehyde, 2-hydroxy-5-methoxy-benzaldehyde, 5-methoxy-2-hydroxybenzaldehyde, 2-hydroxy-5-methoxy benzaldehyde, 2-formyl-4-methoxyphenol, pubchem2649 | |
| 2-hydroxy-5-methoxybenzaldehyde |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 211-589-0
RTECSNumber : BZ2800000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only