missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2'-Hydroxy-4'-methylacetophenone 95.0+%, TCI America™
Supplier: TCI America H153025G
Specifications
| 2′-Hydroxy-4′-methylacetophenone | |
| Yellow | |
| C9H10O2 | |
| 25 g | |
| LYKDOWJROLHYOT-UHFFFAOYSA-N | |
| 1-(2-hydroxy-4-methylphenyl)ethanone | |
| 81338 | |
| ≥95.0% (GC) |
| 6921-64-8 | |
| 245°C | |
| MFCD00100559 | |
| 1-2-hydroxy-4-methylphenyl ethanone, 1-2-hydroxy-4-methylphenyl ethan-1-one, 2'-hydroxy-4'-methylacetophenone, ethanone, 1-2-hydroxy-4-methylphenyl, 2-hydroxy-4-methylacetophenone, 4-methyl-2-hydroxyacetophenone, 1-2-hydroxy-4-methyl-phenyl-ethanone, unii-q168p7rg3b, 1-acetyl-2-hydroxy-4-methylbenzene | |
| CC1=CC(=C(C=C1)C(=O)C)O | |
| 150.177 | |
| 150.18 | |
| Liquid |
Chemical Identifiers
| 6921-64-8 | |
| 150.177 | |
| LYKDOWJROLHYOT-UHFFFAOYSA-N | |
| 81338 | |
| CC1=CC(=C(C=C1)C(=O)C)O |
| C9H10O2 | |
| MFCD00100559 | |
| 1-2-hydroxy-4-methylphenyl ethanone, 1-2-hydroxy-4-methylphenyl ethan-1-one, 2'-hydroxy-4'-methylacetophenone, ethanone, 1-2-hydroxy-4-methylphenyl, 2-hydroxy-4-methylacetophenone, 4-methyl-2-hydroxyacetophenone, 1-2-hydroxy-4-methyl-phenyl-ethanone, unii-q168p7rg3b, 1-acetyl-2-hydroxy-4-methylbenzene | |
| 1-(2-hydroxy-4-methylphenyl)ethanone |
Safety and Handling
TSCA : Yes