accessibility menu, dialog, popup

UserName

CH3CH3OHO

CAS RN 6921-64-8

IUPAC Name: 1-(2-hydroxy-4-methylphenyl)ethan-1-one
Synonyms: 1-(2-hydroxy-4-methylphenyl)ethanone
Molecular Weight (g/mol): 150.18
Molecular Formula: C9H10O2
InChi Key: LYKDOWJROLHYOT-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(C)C=C1O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results