missing translation for 'onlineSavingsMsg'
Learn More

2'-Hydroxy-4'-methylacetophenone 95.0+%, TCI America™

Catalog No. p-7131095
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H15305G 5 g
H153025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H15305G Supplier TCI America Supplier No. H15305G
Only null left

Chemical Identifiers

CAS 6921-64-8
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.177
MDL Number MFCD00100559
InChI Key LYKDOWJROLHYOT-UHFFFAOYSA-N
Synonym 1-2-hydroxy-4-methylphenyl ethanone, 1-2-hydroxy-4-methylphenyl ethan-1-one, 2'-hydroxy-4'-methylacetophenone, ethanone, 1-2-hydroxy-4-methylphenyl, 2-hydroxy-4-methylacetophenone, 4-methyl-2-hydroxyacetophenone, 1-2-hydroxy-4-methyl-phenyl-ethanone, unii-q168p7rg3b, 1-acetyl-2-hydroxy-4-methylbenzene
PubChem CID 81338
IUPAC Name 1-(2-hydroxy-4-methylphenyl)ethanone
SMILES CC1=CC(=C(C=C1)C(=O)C)O

Specifications

Color Yellow
Quantity 5 g
Formula Weight 150.18
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2′-Hydroxy-4′-methylacetophenone