missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-4-methoxybenzophenone-5-sulfonic acid hydrate, tech. 85%, may cont. up to 10% 2-propanol

Catalog No. p-7046042
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1291209 10 g
AAL1291218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1291209 Supplier Thermo Scientific Chemicals Supplier No. L1291209
Only null left

CAS: 4065-45-6 | C14H12O6S | 308.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4065-45-6
Molecular Formula C14H12O6S
Molecular Weight (g/mol) 308.30
MDL Number MFCD00024962
InChI Key CXVGEDCSTKKODG-UHFFFAOYSA-N
Synonym sulisobenzone, benzophenone-4, sungard, 2-hydroxy-4-methoxybenzophenone-5-sulfonic acid, benzophenone 4, uval, uvinul, benzenesulfonic acid, 5-benzoyl-4-hydroxy-2-methoxy, sulisobenzona, sulisobenzonum
PubChem CID 19988
IUPAC Name 5-benzoyl-4-hydroxy-2-methoxybenzenesulfonic acid
SMILES COC1=CC(O)=C(C=C1S(O)(=O)=O)C(=O)C1=CC=CC=C1

Specifications

Melting Point 108°C to 112°C
Quantity 10 g
Beilstein 2889165
Merck Index 14,8983
Formula Weight 308.31 (Anhydrous)
Percent Purity 85%
Grade Technical
Assay may cont. up to 10% 2-propanol
Chemical Name or Material 2-Hydroxy-4-methoxybenzophenone-5-sulfonic acid hydrate
Hazard Category H315-H317-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P501c
EINECSNumber 223-772-2
RTECSNumber DB5044300
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only