missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Hydroxy-4-methoxybenzophenone-5-sulfonic Acid Hydrate (contains 5-10% Isopropyl Alcohol) 98.0+%, TCI America™
$94.33 - $643.00
Chemical Identifiers
| CAS | 4065-45-6 |
|---|---|
| Molecular Formula | C14H12O6S |
| Molecular Weight (g/mol) | 308.30 |
| MDL Number | MFCD00024962 |
| InChI Key | CXVGEDCSTKKODG-UHFFFAOYSA-N |
| Synonym | sulisobenzone, benzophenone-4, sungard, 2-hydroxy-4-methoxybenzophenone-5-sulfonic acid, benzophenone 4, uval, uvinul, benzenesulfonic acid, 5-benzoyl-4-hydroxy-2-methoxy, sulisobenzona, sulisobenzonum |
| PubChem CID | 19988 |
| IUPAC Name | 5-benzoyl-4-hydroxy-2-methoxybenzene-1-sulfonic acid |
| SMILES | COC1=CC(O)=C(C=C1S(O)(=O)=O)C(=O)C1=CC=CC=C1 |
Chemical Identifiers
| 4065-45-6 | |
| 308.30 | |
| CXVGEDCSTKKODG-UHFFFAOYSA-N | |
| 19988 | |
| COC1=CC(O)=C(C=C1S(O)(=O)=O)C(=O)C1=CC=CC=C1 |
| C14H12O6S | |
| MFCD00024962 | |
| sulisobenzone, benzophenone-4, sungard, 2-hydroxy-4-methoxybenzophenone-5-sulfonic acid, benzophenone 4, uval, uvinul, benzenesulfonic acid, 5-benzoyl-4-hydroxy-2-methoxy, sulisobenzona, sulisobenzonum | |
| 5-benzoyl-4-hydroxy-2-methoxybenzene-1-sulfonic acid |
Specifications
| 4065-45-6 | |
| C14H12O6S | |
| 25 g | |
| CXVGEDCSTKKODG-UHFFFAOYSA-N | |
| 5-benzoyl-4-hydroxy-2-methoxybenzene-1-sulfonic acid | |
| 19988 | |
| ≥98.0% (T) | |
| 2-Hydroxy-4-methoxybenzophenone-5-sulfonic Acid Hydrate (contains 5-10% Isopropyl Alcohol) |
| White-Yellow | |
| MFCD00024962 | |
| sulisobenzone, benzophenone-4, sungard, 2-hydroxy-4-methoxybenzophenone-5-sulfonic acid, benzophenone 4, uval, uvinul, benzenesulfonic acid, 5-benzoyl-4-hydroxy-2-methoxy, sulisobenzona, sulisobenzonum | |
| COC1=CC(O)=C(C=C1S(O)(=O)=O)C(=O)C1=CC=CC=C1 | |
| 308.30 | |
| 308.30 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (4)-0145&(4)-0160
RTECSNumber : DB5044300
TSCA : Yes