missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-4-methoxybenzophenone-5-sulfonic Acid Hydrate (contains 5-10% Isopropyl Alcohol) 98.0+%, TCI America™

Catalog No. H046625G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H046625G 25 g
H0466500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H046625G Supplier TCI America Supplier No. H046625G

Chemical Identifiers

CAS 4065-45-6
Molecular Formula C14H12O6S
Molecular Weight (g/mol) 308.30
MDL Number MFCD00024962
InChI Key CXVGEDCSTKKODG-UHFFFAOYSA-N
Synonym sulisobenzone, benzophenone-4, sungard, 2-hydroxy-4-methoxybenzophenone-5-sulfonic acid, benzophenone 4, uval, uvinul, benzenesulfonic acid, 5-benzoyl-4-hydroxy-2-methoxy, sulisobenzona, sulisobenzonum
PubChem CID 19988
IUPAC Name 5-benzoyl-4-hydroxy-2-methoxybenzene-1-sulfonic acid
SMILES COC1=CC(O)=C(C=C1S(O)(=O)=O)C(=O)C1=CC=CC=C1

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 308.30
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxy-4-methoxybenzophenone-5-sulfonic Acid Hydrate (contains 5-10% Isopropyl Alcohol)
EINECSNumber (4)-0145&(4)-0160
RTECSNumber DB5044300
TSCA Yes