missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2'-Hydroxy-4',6'-dimethoxy-2-phenylacetophenone 94.0+%, TCI America™
$305.70 - $913.38
Chemical Identifiers
| CAS | 39604-66-5 |
|---|---|
| Molecular Formula | C16H16O4 |
| Molecular Weight (g/mol) | 272.30 |
| MDL Number | MFCD00218596 |
| InChI Key | NIWDJRBUBVCKAV-UHFFFAOYSA-N |
| Synonym | Benzyl 2-Hydroxy-4,6-dimethoxyphenyl Ketone |
| PubChem CID | 689040 |
| IUPAC Name | 1-(2-hydroxy-4,6-dimethoxyphenyl)-2-phenylethan-1-one |
| SMILES | COC1=CC(O)=C(C(=O)CC2=CC=CC=C2)C(OC)=C1 |
Chemical Identifiers
| 39604-66-5 | |
| 272.30 | |
| NIWDJRBUBVCKAV-UHFFFAOYSA-N | |
| 689040 | |
| COC1=CC(O)=C(C(=O)CC2=CC=CC=C2)C(OC)=C1 |
| C16H16O4 | |
| MFCD00218596 | |
| Benzyl 2-Hydroxy-4,6-dimethoxyphenyl Ketone | |
| 1-(2-hydroxy-4,6-dimethoxyphenyl)-2-phenylethan-1-one |
Specifications
| 39604-66-5 | |
| White-Yellow | |
| MFCD00218596 | |
| Benzyl 2-Hydroxy-4,6-dimethoxyphenyl Ketone | |
| COC1=CC(O)=C(C(=O)CC2=CC=CC=C2)C(OC)=C1 | |
| 272.30 | |
| 272.30 | |
| Crystalline Powder |
| 117°C | |
| C16H16O4 | |
| 1 g | |
| NIWDJRBUBVCKAV-UHFFFAOYSA-N | |
| 1-(2-hydroxy-4,6-dimethoxyphenyl)-2-phenylethan-1-one | |
| 689040 | |
| ≥94.0% (GC) | |
| 2′-Hydroxy-4′,6′-dimethoxy-2-phenylacetophenone |