missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2'-Hydroxy-4',6'-dimethoxy-2-phenylacetophenone 94.0+%, TCI America™
Supplier: TCI America H15775G
Specifications
| 2′-Hydroxy-4′,6′-dimethoxy-2-phenylacetophenone | |
| 117°C | |
| C16H16O4 | |
| 5 g | |
| NIWDJRBUBVCKAV-UHFFFAOYSA-N | |
| 1-(2-hydroxy-4,6-dimethoxyphenyl)-2-phenylethan-1-one | |
| 689040 | |
| ≥94.0% (GC) |
| 39604-66-5 | |
| White-Yellow | |
| MFCD00218596 | |
| Benzyl 2-Hydroxy-4,6-dimethoxyphenyl Ketone | |
| COC1=CC(O)=C(C(=O)CC2=CC=CC=C2)C(OC)=C1 | |
| 272.30 | |
| 272.30 | |
| Crystalline Powder |
Chemical Identifiers
| 39604-66-5 | |
| 272.30 | |
| NIWDJRBUBVCKAV-UHFFFAOYSA-N | |
| 689040 | |
| COC1=CC(O)=C(C(=O)CC2=CC=CC=C2)C(OC)=C1 |
| C16H16O4 | |
| MFCD00218596 | |
| Benzyl 2-Hydroxy-4,6-dimethoxyphenyl Ketone | |
| 1-(2-hydroxy-4,6-dimethoxyphenyl)-2-phenylethan-1-one |