missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-3-methoxybenzyl Alcohol 98.0+%, TCI America™

Catalog No. p-7130953
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H12731G 1 g
H12735G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H12731G Supplier TCI America Supplier No. H12731G

Chemical Identifiers

CAS 4383-05-5
Molecular Formula C8H10O3
Molecular Weight (g/mol) 154.17
MDL Number MFCD00008362
InChI Key OSZHSESNQIMXMZ-UHFFFAOYSA-N
Synonym 2-(Hydroxymethyl)-6-methoxyphenol, 2-Hydroxy-3-(hydroxymethyl)anisole
PubChem CID 78088
IUPAC Name 2-(hydroxymethyl)-6-methoxyphenol
SMILES COC1=CC=CC(=C1O)CO

Specifications

Melting Point 63°C
Color White-Yellow
Quantity 1 g
Formula Weight 154.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxy-3-methoxybenzyl Alcohol