accessibility menu, dialog, popup

UserName

CH3OOHHO

CAS RN 4383-05-5

IUPAC Name: 2-(hydroxymethyl)-6-methoxyphenol
Synonyms: 2-(hydroxymethyl)-6-methoxyphenol
Molecular Weight (g/mol): 154.17
Molecular Formula: C8H10O3
InChi Key: OSZHSESNQIMXMZ-UHFFFAOYSA-N
SMILES: COC1=CC=CC(CO)=C1O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results