missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-3-methoxy-5-nitrobenzaldehyde, 98%

Catalog No. AAA1436003
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1436003 1 g
AAA1436006 5 g
AAA1436014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1436003 Supplier Thermo Scientific Chemicals Supplier No. A1436003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 17028-61-4 | C8H7NO5 | 197.15 g/mol

2-Hydroxy-3-methoxy-5-nitrobenzaldehyde is used for the syntheses of zinc-selective spiropyran-based fluorescent and photoregenerable receptor, (E)-2,4-dihydroxy-N?-(2-hydroxy-3-methoxy-5-nitrobenzylidene)benzohydrazide dihydrate, ethyl 4-(8-methoxy-3?,3?-dimethyl-6-nitrospiro[chromene-2,2?-indoline]-1?-yl)butanoate and 1-(3-carbomethoxypropyl)-3-3-dimethyl-8-methoxy-6-nitrospiro[2H-1]-benzopyran-2,2-indoline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hydroxy-3-methoxy-5-nitrobenzaldehyde is used for the syntheses of zinc-selective spiropyran-based fluorescent and photoregenerable receptor, (E)-2,4-dihydroxy-N′-(2-hydroxy-3-methoxy-5-nitrobenzylidene)benzohydrazide dihydrate, ethyl 4-(8-methoxy-3′,3′-dimethyl-6-nitrospiro[chromene-2,2′-indoline]-1′-yl)butanoate and 1-(3-carbomethoxypropyl)-3-3-dimethyl-8-methoxy-6-nitrospiro[2H-1]-benzopyran-2,2-indoline.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents. It is sensitive to air.

Chemical Identifiers

CAS 17028-61-4
Molecular Formula C8H7NO5
Molecular Weight (g/mol) 197.15
MDL Number MFCD00017033
InChI Key HGKHVFKBOHFYSS-UHFFFAOYSA-N
PubChem CID 307886
IUPAC Name 2-hydroxy-3-methoxy-5-nitrobenzaldehyde
SMILES COC1=CC(=CC(C=O)=C1O)[N+]([O-])=O

Specifications

Melting Point 138°C to 142°C
Quantity 1 g
Beilstein 613271
Sensitivity Air Sensitive
Synonym 3-methoxy-5-nitrosalicylaldehyde, 2-hydroxy-5-nitro-m-anisaldehyde, benzaldehyde, 2-hydroxy-3-methoxy-5-nitro, 5-nitro-o-vanillin, acmc-1bt2m, 3-methoxy-5-nitrosalicyladehyde, 2-hydroxy-5-nitro-3-methoxybenzaldehyde, benzaldehyde,2-hydroxy-3-methoxy-5-nitro
Solubility Information Slightly soluble in water.
Formula Weight 197.15
Percent Purity 98%
Chemical Name or Material 2-Hydroxy-3-methoxy-5-nitrobenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only