missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Hydroxy-2-propanesulfonic acid monosodium salt, 97%
CAS: 540-92-1 | C3H8NaO4S+ | 163.143 g/mol
$169.47 - $674.34
Chemical Identifiers
| CAS | 540-92-1 |
|---|---|
| Molecular Formula | C3H8NaO4S+ |
| Molecular Weight (g/mol) | 163.143 |
| MDL Number | MFCD00031395 |
| InChI Key | YNJORDSKPXMABC-UHFFFAOYSA-N |
| Synonym | sodium acetone bisulfite, acetone-sodium bisulfite adduct, sodium bisulfite-acetone adduct, acetone-monosodium sulfite adduct, sodium 2-hydroxy-2-propane-sulfonate, acetone-sodium bisulfite compound 1:1, acetone, compound with sodium bisulfite 1:1, 2-propanone, compd. with monosodium sulfite 1:1, 2-propanesulfonic acid, monosodium salt, sodium 2-hydroxypropane-2-sulfonic acid |
| PubChem CID | 54602011 |
| IUPAC Name | sodium;2-hydroxypropane-2-sulfonic acid |
| SMILES | CC(C)(O)S(=O)(=O)O.[Na+] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 540-92-1 | |
| 163.143 | |
| YNJORDSKPXMABC-UHFFFAOYSA-N | |
| 54602011 | |
| CC(C)(O)S(=O)(=O)O.[Na+] |
| C3H8NaO4S+ | |
| MFCD00031395 | |
| sodium acetone bisulfite, acetone-sodium bisulfite adduct, sodium bisulfite-acetone adduct, acetone-monosodium sulfite adduct, sodium 2-hydroxy-2-propane-sulfonate, acetone-sodium bisulfite compound 1:1, acetone, compound with sodium bisulfite 1:1, 2-propanone, compd. with monosodium sulfite 1:1, 2-propanesulfonic acid, monosodium salt, sodium 2-hydroxypropane-2-sulfonic acid | |
| sodium;2-hydroxypropane-2-sulfonic acid |
Specifications
| 540-92-1 | |
| MFCD00031395 | |
| sodium acetone bisulfite, acetone-sodium bisulfite adduct, sodium bisulfite-acetone adduct, acetone-monosodium sulfite adduct, sodium 2-hydroxy-2-propane-sulfonate, acetone-sodium bisulfite compound 1:1, acetone, compound with sodium bisulfite 1:1, 2-propanone, compd. with monosodium sulfite 1:1, 2-propanesulfonic acid, monosodium salt, sodium 2-hydroxypropane-2-sulfonic acid | |
| CC(C)(O)S(=O)(=O)O.[Na+] | |
| 163.143 | |
| 162.14 | |
| Crystalline |
| C3H8NaO4S+ | |
| 100 g | |
| YNJORDSKPXMABC-UHFFFAOYSA-N | |
| sodium;2-hydroxypropane-2-sulfonic acid | |
| 54602011 | |
| 97% | |
| 2-Hydroxy-2-propanesulfonic acid monosodium salt |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 208-761-2
RTECSNumber : TZ6550000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only