missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-2-propanesulfonic acid monosodium salt, 97%

CAS: 540-92-1 | C3H8NaO4S+ | 163.143 g/mol

Supplier:  Thermo Scientific Chemicals 03625622

 View more versions of this product

Catalog No. AA3625622


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Hydroxy-2-propanesulfonic acid monosodium salt
C3H8NaO4S+
100 g
YNJORDSKPXMABC-UHFFFAOYSA-N
sodium;2-hydroxypropane-2-sulfonic acid
54602011
97%
540-92-1
MFCD00031395
sodium acetone bisulfite, acetone-sodium bisulfite adduct, sodium bisulfite-acetone adduct, acetone-monosodium sulfite adduct, sodium 2-hydroxy-2-propane-sulfonate, acetone-sodium bisulfite compound 1:1, acetone, compound with sodium bisulfite 1:1, 2-propanone, compd. with monosodium sulfite 1:1, 2-propanesulfonic acid, monosodium salt, sodium 2-hydroxypropane-2-sulfonic acid
CC(C)(O)S(=O)(=O)O.[Na+]
163.143
162.14
Crystalline

Chemical Identifiers

540-92-1
163.143
YNJORDSKPXMABC-UHFFFAOYSA-N
54602011
CC(C)(O)S(=O)(=O)O.[Na+]
C3H8NaO4S+
MFCD00031395
sodium acetone bisulfite, acetone-sodium bisulfite adduct, sodium bisulfite-acetone adduct, acetone-monosodium sulfite adduct, sodium 2-hydroxy-2-propane-sulfonate, acetone-sodium bisulfite compound 1:1, acetone, compound with sodium bisulfite 1:1, 2-propanone, compd. with monosodium sulfite 1:1, 2-propanesulfonic acid, monosodium salt, sodium 2-hydroxypropane-2-sulfonic acid
sodium;2-hydroxypropane-2-sulfonic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 208-761-2

RTECSNumber : TZ6550000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only