CAS RN 540-92-1
CAS RN 540-92-1
IUPAC Name:
2-hydroxypropane-2-sulfonic acid sodium
Synonyms:
2-hydroxypropane-2-sulfonic acid sodium
Molecular Weight (g/mol):
163.14
Molecular Formula:
C3H8NaO4S
InChi Key:
RJQNNQWGSIPDML-UHFFFAOYSA-N
SMILES:
[Na].CC(C)(O)S(O)(=O)=O
1
–
1
of
1
results
1
2-Hydroxy-2-propanesulfonic acid monosodium salt, 97%
CAS: 540-92-1 Molecular Formula: C3H8NaO4S+ Molecular Weight (g/mol): 163.143 MDL Number: MFCD00031395 InChI Key: YNJORDSKPXMABC-UHFFFAOYSA-N Synonym: sodium acetone bisulfite,acetone-sodium bisulfite adduct,sodium bisulfite-acetone adduct,acetone-monosodium sulfite adduct,sodium 2-hydroxy-2-propane-sulfonate,acetone-sodium bisulfite compound 1:1,acetone, compound with sodium bisulfite 1:1,2-propanone, compd. with monosodium sulfite 1:1,2-propanesulfonic acid, monosodium salt,sodium 2-hydroxypropane-2-sulfonic acid PubChem CID: 54602011 IUPAC Name: sodium;2-hydroxypropane-2-sulfonic acid SMILES: CC(C)(O)S(=O)(=O)O.[Na+]