missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Hexylthiophene 98.0+%, TCI America™
$96.26 - $321.51
Chemical Identifiers
| CAS | 18794-77-9 |
|---|---|
| Molecular Formula | C10H16S |
| Molecular Weight (g/mol) | 168.30 |
| MDL Number | MFCD00022535 |
| InChI Key | QZVHYFUVMQIGGM-UHFFFAOYSA-N |
| Synonym | thiophene, 2-hexyl, 2-n-hexylthiophene, 2-hexyl-thiophene, hexylthiophene, thiophene, hexyl, 2-hexyl thiophene, 2-? hexylthiophene, 2-hexylthiophene, ksc179k7d |
| PubChem CID | 87793 |
| IUPAC Name | 2-hexylthiophene |
| SMILES | CCCCCCC1=CC=CS1 |
Chemical Identifiers
| 18794-77-9 | |
| 168.30 | |
| QZVHYFUVMQIGGM-UHFFFAOYSA-N | |
| 87793 | |
| CCCCCCC1=CC=CS1 |
| C10H16S | |
| MFCD00022535 | |
| thiophene, 2-hexyl, 2-n-hexylthiophene, 2-hexyl-thiophene, hexylthiophene, thiophene, hexyl, 2-hexyl thiophene, 2-? hexylthiophene, 2-hexylthiophene, ksc179k7d | |
| 2-hexylthiophene |
Specifications
| 18794-77-9 | |
| 92°C | |
| MFCD00022535 | |
| thiophene, 2-hexyl, 2-n-hexylthiophene, 2-hexyl-thiophene, hexylthiophene, thiophene, hexyl, 2-hexyl thiophene, 2-? hexylthiophene, 2-hexylthiophene, ksc179k7d | |
| CCCCCCC1=CC=CS1 | |
| 168.30 | |
| 168.30 | |
| Liquid |
| Yellow | |
| C10H16S | |
| 5 g | |
| QZVHYFUVMQIGGM-UHFFFAOYSA-N | |
| 2-hexylthiophene | |
| 87793 | |
| ≥98.0% (GC) | |
| 2-Hexylthiophene |
Safety and Handling
TSCA : No