missing translation for 'onlineSavingsMsg'
Learn More

2-Hexylthiophene 98.0+%, TCI America™

Catalog No. h13505g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
H13505G 5 g
H135025G 25 g
2 options

Catalog No. H13505G

Supplier: TCI America H13505G

Only null left

Chemical Identifiers

CAS 18794-77-9
Molecular Formula C10H16S
Molecular Weight (g/mol) 168.30
MDL Number MFCD00022535
InChI Key QZVHYFUVMQIGGM-UHFFFAOYSA-N
Synonym thiophene, 2-hexyl, 2-n-hexylthiophene, 2-hexyl-thiophene, hexylthiophene, thiophene, hexyl, 2-hexyl thiophene, 2-? hexylthiophene, 2-hexylthiophene, ksc179k7d
PubChem CID 87793
IUPAC Name 2-hexylthiophene
SMILES CCCCCCC1=CC=CS1

Specifications

Color Yellow
Boiling Point 92°C
Quantity 5 g
Formula Weight 168.30
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Hexylthiophene