accessibility menu, dialog, popup

UserName

H3CS

CAS RN 18794-77-9

IUPAC Name: 2-hexylthiophene
Synonyms: 2-hexylthiophene
Molecular Weight (g/mol): 168.3
Molecular Formula: C10H16S
InChi Key: QZVHYFUVMQIGGM-UHFFFAOYSA-N
SMILES: CCCCCCC1=CC=CS1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)