missing translation for 'onlineSavingsMsg'
Learn More

2-Hexyldecanoic Acid 98.0+%, TCI America™

Supplier:  TCI America H0507500ML

 View more versions of this product

Catalog No. H0507500ML


Only null left

Description

Description

Specifications

Specifications

2-Hexyldecanoic Acid
Yellow
C16H32O2
500 mL
JMOLZNNXZPAGBH-UHFFFAOYSA-N
2-hexyldecanoic acid
32912
≥98.0% (GC)
25354-97-6
150°C
MFCD00059651
decanoic acid, 2-hexyl, hexyldecanoic acid, c16 guerbet fatty acid, 2-hexyldecansaeure, 7-pentadecanecarboxylic acid, 2-hexyldecansaeure german, 2-hexyl-decanoic acid, 2-n-hexyldecanoic acid, 2-hexyldecanoicacid, isocarb 16
CCCCCCCCC(CCCCCC)C(=O)O
256.43
256.43
Liquid

Chemical Identifiers

25354-97-6
256.43
JMOLZNNXZPAGBH-UHFFFAOYSA-N
32912
CCCCCCCCC(CCCCCC)C(=O)O
C16H32O2
MFCD00059651
decanoic acid, 2-hexyl, hexyldecanoic acid, c16 guerbet fatty acid, 2-hexyldecansaeure, 7-pentadecanecarboxylic acid, 2-hexyldecansaeure german, 2-hexyl-decanoic acid, 2-n-hexyldecanoic acid, 2-hexyldecanoicacid, isocarb 16
2-hexyldecanoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0608

RTECSNumber : HD9750000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions