missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Hexyl-1-decanol 97.0+%, TCI America™
$66.46 - $558.20
Chemical Identifiers
| CAS | 2425-77-6 |
|---|---|
| Molecular Formula | C16H34O |
| Molecular Weight (g/mol) | 242.447 |
| MDL Number | MFCD00060903 |
| InChI Key | XULHFMYCBKQGEE-UHFFFAOYSA-N |
| Synonym | 2-Hexyldecyl Alcohol |
| PubChem CID | 95337 |
| IUPAC Name | 2-hexyldecan-1-ol |
| SMILES | CCCCCCCCC(CCCCCC)CO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
H146125ML
|
N/A | 25 mL |
Each for $66.46
|
|
|||||
|
H1461100ML
|
N/A | 100 mL |
Each for $210.75
|
|
|||||
|
H1461500ML
|
N/A | 500 mL |
Each for $558.20
|
|
|||||
Chemical Identifiers
| C16H34O | |
| MFCD00060903 | |
| 2-Hexyldecyl Alcohol | |
| 2-hexyldecan-1-ol |
Specifications
| 2425-77-6 | |
| Colorless | |
| C16H34O | |
| 2-Hexyldecyl Alcohol | |
| CCCCCCCCC(CCCCCC)CO | |
| 242.447 | |
| 242.45 | |
| Liquid |
| -15°C | |
| 107°C | |
| MFCD00060903 | |
| XULHFMYCBKQGEE-UHFFFAOYSA-N | |
| 2-hexyldecan-1-ol | |
| 95337 | |
| ≥97.0% (GC) | |
| 2-Hexyl-1-decanol |
Safety and Handling
EINECSNumber : (2)-0217
TSCA : Yes