missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Hexyl-1-decanol 97.0+%, TCI America™
Supplier: TCI America H146125ML
| Quantity | 25 mL |
|---|
Chemical Identifiers
| C16H34O | |
| MFCD00060903 | |
| 2-Hexyldecyl Alcohol | |
| 2-hexyldecan-1-ol |
Specifications
| 2-Hexyl-1-decanol | |
| -15°C | |
| 107°C | |
| 25 mL | |
| 2-Hexyldecyl Alcohol | |
| CCCCCCCCC(CCCCCC)CO | |
| 242.447 | |
| 242.45 | |
| Liquid |
| 2425-77-6 | |
| Colorless | |
| C16H34O | |
| MFCD00060903 | |
| XULHFMYCBKQGEE-UHFFFAOYSA-N | |
| 2-hexyldecan-1-ol | |
| 95337 | |
| ≥97.0% (GC) |
Safety and Handling
EINECSNumber : (2)-0217
TSCA : Yes