missing translation for 'onlineSavingsMsg'
Learn More

2-Hexyl-1-decanol 97.0+%, TCI America™

Supplier:  TCI America H1461500ML

Quantity 500 mL

 View more versions of this product

Catalog No. H1461500ML


Only null left

Description

Description

Specifications

Chemical Identifiers

2425-77-6
242.447
XULHFMYCBKQGEE-UHFFFAOYSA-N
95337
CCCCCCCCC(CCCCCC)CO
C16H34O
MFCD00060903
2-Hexyldecyl Alcohol
2-hexyldecan-1-ol

Specifications

2-Hexyl-1-decanol
-15°C
107°C
500 mL
2-Hexyldecyl Alcohol
CCCCCCCCC(CCCCCC)CO
242.447
242.45
Liquid
2425-77-6
Colorless
C16H34O
MFCD00060903
XULHFMYCBKQGEE-UHFFFAOYSA-N
2-hexyldecan-1-ol
95337
≥97.0% (GC)
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0217

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions