missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| 2-Hexanol | |
| Colorless | |
| C6H14O | |
| 25 mL | |
| 2-hexanol, sec-hexyl alcohol, 2-hydroxyhexane, 2-hexyl alcohol, n-butylmethylcarbinol, 2-hexanol, r, 2-hexanol, s, n-c4h9ch oh ch3, methylamyl alcohol, sec-hexanol | |
| CCCCC(C)O | |
| 102.177 | |
| 102.18 | |
| Liquid |
| 626-93-7 | |
| 139°C | |
| MFCD00004585 | |
| 2282 | |
| QNVRIHYSUZMSGM-UHFFFAOYSA-N | |
| hexan-2-ol | |
| 12297 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 626-93-7 | |
| 102.177 | |
| QNVRIHYSUZMSGM-UHFFFAOYSA-N | |
| 12297 | |
| CCCCC(C)O |
| C6H14O | |
| MFCD00004585 | |
| 2-hexanol, sec-hexyl alcohol, 2-hydroxyhexane, 2-hexyl alcohol, n-butylmethylcarbinol, 2-hexanol, r, 2-hexanol, s, n-c4h9ch oh ch3, methylamyl alcohol, sec-hexanol | |
| hexan-2-ol |
Safety and Handling
EINECSNumber : (2)-0217
RTECSNumber : MO8470000
TSCA : Yes