missing translation for 'onlineSavingsMsg'
Learn More

2-Hexanol 98.0+%, TCI America™

Catalog No. p-7130360
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H013125ML 25 mL
H0131100ML 100 mL
H0131500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H013125ML Supplier TCI America Supplier No. H013125ML

Chemical Identifiers

CAS 626-93-7
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.177
MDL Number MFCD00004585
InChI Key QNVRIHYSUZMSGM-UHFFFAOYSA-N
Synonym 2-hexanol, sec-hexyl alcohol, 2-hydroxyhexane, 2-hexyl alcohol, n-butylmethylcarbinol, 2-hexanol, r, 2-hexanol, s, n-c4h9ch oh ch3, methylamyl alcohol, sec-hexanol
PubChem CID 12297
IUPAC Name hexan-2-ol
SMILES CCCCC(C)O

Specifications

Color Colorless
Boiling Point 139°C
Quantity 25 mL
UN Number 2282
Formula Weight 102.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Hexanol
EINECSNumber (2)-0217
RTECSNumber MO8470000
TSCA Yes