missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Heptene (cis- and trans- mixture) 98.0+%, TCI America™
Supplier: TCI America H004325ML
Specifications
| 2-Heptene (cis- and trans- mixture) | |
| Colorless | |
| C7H14 | |
| 25 mL | |
| OTTZHAVKAVGASB-HWKANZROSA-N | |
| (E)-hept-2-ene | |
| 639662 | |
| ≥98.0% (GC) |
| 592-77-8 | |
| 98°C | |
| MFCD00009506 | |
| 2278 | |
| CCCCC=CC | |
| 98.189 | |
| 98.19 | |
| Liquid |
Chemical Identifiers
| 592-77-8 | |
| 98.189 | |
| OTTZHAVKAVGASB-HWKANZROSA-N | |
| (E)-hept-2-ene |
| C7H14 | |
| MFCD00009506 | |
| 639662 | |
| CCCCC=CC |
Safety and Handling
TSCA : No