accessibility menu, dialog, popup

UserName

CAS RN 592-77-8

IUPAC Name: (2E)-hept-2-ene
Synonyms: 2-heptene (trans)
Molecular Weight (g/mol): 98.19
Molecular Formula: C7H14
InChi Key: OTTZHAVKAVGASB-HWKANZROSA-N
SMILES: CCCC\C=C\C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results