missing translation for 'onlineSavingsMsg'
Learn More

2-Heptene (cis- and trans- mixture) 98.0+%, TCI America™

Catalog No. p-7130307
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H00435ML 5 mL
H004325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H00435ML Supplier TCI America Supplier No. H00435ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 592-77-8
Molecular Formula C7H14
Molecular Weight (g/mol) 98.189
MDL Number MFCD00009506
InChI Key OTTZHAVKAVGASB-HWKANZROSA-N
PubChem CID 639662
IUPAC Name (E)-hept-2-ene
SMILES CCCCC=CC

Specifications

Color Colorless
Boiling Point 98°C
Quantity 5 mL
UN Number 2278
Formula Weight 98.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Heptene (cis- and trans- mixture)