missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Heptanol 98.0+%, TCI America™
$51.26 - $210.75
Chemical Identifiers
| CAS | 543-49-7 |
|---|---|
| Molecular Formula | C7H16O |
| Molecular Weight (g/mol) | 116.20 |
| MDL Number | MFCD00004587 |
| InChI Key | CETWDUZRCINIHU-UHFFFAOYNA-N |
| Synonym | 2-heptanol, s-heptyl alcohol, 2-heptyl alcohol, 2-hydroxyheptane, amyl methyl carbinol, heptanol-2, methyl amyl carbinol, 1-methylhexanol, heptyl alcohol, sec, 2-heptanol natural |
| PubChem CID | 10976 |
| IUPAC Name | heptan-2-ol |
| SMILES | CCCCCC(C)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
H003425ML
|
N/A | 25 mL |
Each for $51.26
|
|
|||||
|
H0034100ML
|
N/A | 100 mL |
Each for $81.64
|
|
|||||
|
H0034500ML
|
N/A | 500 mL |
Each for $210.75
|
|
|||||
Chemical Identifiers
| C7H16O | |
| MFCD00004587 | |
| 2-heptanol, s-heptyl alcohol, 2-heptyl alcohol, 2-hydroxyheptane, amyl methyl carbinol, heptanol-2, methyl amyl carbinol, 1-methylhexanol, heptyl alcohol, sec, 2-heptanol natural | |
| heptan-2-ol |
Specifications
| 543-49-7 | |
| 160°C | |
| MFCD00004587 | |
| CETWDUZRCINIHU-UHFFFAOYNA-N | |
| heptan-2-ol | |
| 10976 | |
| ≥98.0% (GC) | |
| 2-Heptanol |
| Colorless | |
| C7H16O | |
| 2-heptanol, s-heptyl alcohol, 2-heptyl alcohol, 2-hydroxyheptane, amyl methyl carbinol, heptanol-2, methyl amyl carbinol, 1-methylhexanol, heptyl alcohol, sec, 2-heptanol natural | |
| CCCCCC(C)O | |
| 116.20 | |
| 116.20 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0217
RTECSNumber : MJ2975000
TSCA : Yes