missing translation for 'onlineSavingsMsg'
Learn More

2-Heptanol 98.0+%, TCI America™

Catalog No. H003425ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H003425ML 25 mL
H0034100ML 100 mL
H0034500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H003425ML Supplier TCI America Supplier No. H003425ML

Chemical Identifiers

CAS 543-49-7
Molecular Formula C7H16O
Molecular Weight (g/mol) 116.20
MDL Number MFCD00004587
InChI Key CETWDUZRCINIHU-UHFFFAOYNA-N
Synonym 2-heptanol, s-heptyl alcohol, 2-heptyl alcohol, 2-hydroxyheptane, amyl methyl carbinol, heptanol-2, methyl amyl carbinol, 1-methylhexanol, heptyl alcohol, sec, 2-heptanol natural
PubChem CID 10976
IUPAC Name heptan-2-ol
SMILES CCCCCC(C)O

Specifications

Color Colorless
Boiling Point 160°C
Quantity 25 mL
Formula Weight 116.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Heptanol
EINECSNumber (2)-0217
RTECSNumber MJ2975000
TSCA Yes