missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Heptanol 98.0+%, TCI America™
Supplier: TCI America H0034100ML
| Quantity | 100 mL |
|---|
Chemical Identifiers
| C7H16O | |
| MFCD00004587 | |
| 2-heptanol, s-heptyl alcohol, 2-heptyl alcohol, 2-hydroxyheptane, amyl methyl carbinol, heptanol-2, methyl amyl carbinol, 1-methylhexanol, heptyl alcohol, sec, 2-heptanol natural | |
| heptan-2-ol |
Specifications
| 2-Heptanol | |
| Colorless | |
| C7H16O | |
| MFCD00004587 | |
| CETWDUZRCINIHU-UHFFFAOYNA-N | |
| heptan-2-ol | |
| 10976 | |
| ≥98.0% (GC) |
| 543-49-7 | |
| 160°C | |
| 100 mL | |
| 2-heptanol, s-heptyl alcohol, 2-heptyl alcohol, 2-hydroxyheptane, amyl methyl carbinol, heptanol-2, methyl amyl carbinol, 1-methylhexanol, heptyl alcohol, sec, 2-heptanol natural | |
| CCCCCC(C)O | |
| 116.20 | |
| 116.20 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0217
RTECSNumber : MJ2975000
TSCA : Yes