missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Formylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$55.92 - $55.92
Specifications
| CAS | 40138-16-7 |
|---|---|
| Melting Point | 125°C |
| Molecular Formula | C7H7BO3 |
| MDL Number | MFCD00151822 |
| InChI Key | DGUWACLYDSWXRZ-UHFFFAOYSA-N |
Specifications
| 40138-16-7 | |
| C7H7BO3 | |
| DGUWACLYDSWXRZ-UHFFFAOYSA-N | |
| 149.94 | |
| 149.94 |
| 125°C | |
| MFCD00151822 | |
| (2-formylphenyl)boronic acid | |
| 292189 | |
| Crystalline Powder |