missing translation for 'onlineSavingsMsg'
Learn More

2-(Fmoc-amino)benzoic acid, 98%

CAS: 150256-42-1 | C22H17NO4 | 359.381 g/mol

$111.74 - $457.22

Chemical Identifiers

CAS 150256-42-1
Molecular Formula C22H17NO4
Molecular Weight (g/mol) 359.381
MDL Number MFCD00235883
InChI Key CNAVPEPPAVHHKN-UHFFFAOYSA-N
Synonym fmoc-2-abz-oh, fmoc-2-aminobenzoic acid, 2-fmoc-amino benzoic acid, fmoc-abz-oh, n-fmoc-anthranilic acid, 2-9h-fluoren-9-yl methoxy carbonyl amino benzoic acid, fmoc-anthranilic acid, benzoic acid, 2-9h-fluoren-9-ylmethoxy carbonyl amino, 2-9h-fluoren-9-ylmethoxy carbonyl amino benzoic acid, n-2-9-fluorenylmethyloxycarbonyl amino-benzoic acid
PubChem CID 978386
IUPAC Name 2-(9H-fluoren-9-ylmethoxycarbonylamino)benzoic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC4=CC=CC=C4C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6345906
View Documents
Thermo Scientific Chemicals
H6345906
5 g
Each for $111.74
Only null left
 
AAH6345914
View Documents
Thermo Scientific Chemicals
H6345914
25 g
Each for $457.22
Only null left
 
Description

Description

It is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field

Solubility
Soluble in water or 1% acetic acid.

Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Chemical Identifiers

150256-42-1
359.381
CNAVPEPPAVHHKN-UHFFFAOYSA-N
978386
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC4=CC=CC=C4C(=O)O
C22H17NO4
MFCD00235883
fmoc-2-abz-oh, fmoc-2-aminobenzoic acid, 2-fmoc-amino benzoic acid, fmoc-abz-oh, n-fmoc-anthranilic acid, 2-9h-fluoren-9-yl methoxy carbonyl amino benzoic acid, fmoc-anthranilic acid, benzoic acid, 2-9h-fluoren-9-ylmethoxy carbonyl amino, 2-9h-fluoren-9-ylmethoxy carbonyl amino benzoic acid, n-2-9-fluorenylmethyloxycarbonyl amino-benzoic acid
2-(9H-fluoren-9-ylmethoxycarbonylamino)benzoic acid

Specifications

150256-42-1
White
MFCD00235883
7659726
Soluble in water or 1% acetic acid.
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC4=CC=CC=C4C(=O)O
359.381
359.37
Powder
216°C to 218°C (decomposition)
C22H17NO4
5 g
fmoc-2-abz-oh, fmoc-2-aminobenzoic acid, 2-fmoc-amino benzoic acid, fmoc-abz-oh, n-fmoc-anthranilic acid, 2-9h-fluoren-9-yl methoxy carbonyl amino benzoic acid, fmoc-anthranilic acid, benzoic acid, 2-9h-fluoren-9-ylmethoxy carbonyl amino, 2-9h-fluoren-9-ylmethoxy carbonyl amino benzoic acid, n-2-9-fluorenylmethyloxycarbonyl amino-benzoic acid
CNAVPEPPAVHHKN-UHFFFAOYSA-N
2-(9H-fluoren-9-ylmethoxycarbonylamino)benzoic acid
978386
98%
2-(Fmoc-amino)benzoic acid
Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only