Learn More
2-Fluorophenylhydrazine hydrochloride, 98%
CAS: 2924-15-4 | C6H8ClFN2 | 162.592 g/mol
Supplier: Thermo Scientific Chemicals A1371606
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 2-Fluorophenylhydrazine hydrochloride | |
| ∼205°C (decomposition) | |
| MFCD00012927 | |
| 3627985 | |
| VFZYLSYYMHFPSY-UHFFFAOYSA-N | |
| (2-fluorophenyl)hydrazine;hydrochloride | |
| 2723910 | |
| 98% |
| 2924-15-4 | |
| C6H8ClFN2 | |
| 5 g | |
| 2-fluorophenylhydrazine hydrochloride, 2-fluorophenyl hydrazine hydrochloride, 2-fluorophenylhydrazine hcl, 2-fluorophenylhydrazinehydrochloride, o-fluorophenylhydrazine hydrochloride, hydrazine, 2-fluorophenyl-, monohydrochloride, 1-2-fluorophenyl hydrazine hydrochloride, pubchem3347, c6h7fn2.hcl, acmc-209h7m | |
| C1=CC=C(C(=C1)NN)F.Cl | |
| 162.592 | |
| 162.6 |
Chemical Identifiers
| 2924-15-4 | |
| 162.592 | |
| VFZYLSYYMHFPSY-UHFFFAOYSA-N | |
| 2723910 | |
| C1=CC=C(C(=C1)NN)F.Cl |
| C6H8ClFN2 | |
| MFCD00012927 | |
| 2-fluorophenylhydrazine hydrochloride, 2-fluorophenyl hydrazine hydrochloride, 2-fluorophenylhydrazine hcl, 2-fluorophenylhydrazinehydrochloride, o-fluorophenylhydrazine hydrochloride, hydrazine, 2-fluorophenyl-, monohydrochloride, 1-2-fluorophenyl hydrazine hydrochloride, pubchem3347, c6h7fn2.hcl, acmc-209h7m | |
| (2-fluorophenyl)hydrazine;hydrochloride |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P201-P202-P261-P264b-P270-P271-P272-P280-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P311-P312-P330-P333+P313-P361-P363-P501c
H301+H311+H331-H315-H317-H319-H335-H351
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 220-885-9
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only