missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorophenylhydrazine hydrochloride, 98%

Catalog No. aaa1371603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1371603 1 g
AAA1371606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1371603 Supplier Thermo Scientific Chemicals Supplier No. A1371603
Only null left

CAS: 2924-15-4 | C6H8ClFN2 | 162.592 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2924-15-4
Molecular Formula C6H8ClFN2
Molecular Weight (g/mol) 162.592
MDL Number MFCD00012927
InChI Key VFZYLSYYMHFPSY-UHFFFAOYSA-N
Synonym 2-fluorophenylhydrazine hydrochloride, 2-fluorophenyl hydrazine hydrochloride, 2-fluorophenylhydrazine hcl, 2-fluorophenylhydrazinehydrochloride, o-fluorophenylhydrazine hydrochloride, hydrazine, 2-fluorophenyl-, monohydrochloride, 1-2-fluorophenyl hydrazine hydrochloride, pubchem3347, c6h7fn2.hcl, acmc-209h7m
PubChem CID 2723910
IUPAC Name (2-fluorophenyl)hydrazine;hydrochloride
SMILES C1=CC=C(C(=C1)NN)F.Cl

Specifications

Melting Point ∼205°C (decomposition)
Quantity 1 g
Beilstein 3627985
Formula Weight 162.6
Percent Purity 98%
Chemical Name or Material 2-Fluorophenylhydrazine hydrochloride
Hazard Category H301+H311+H331-H315-H317-H319-H335-H351
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P272-P280-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P311-P312-P330-P333+P313-P361-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 220-885-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only