missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluorophenylacetone 97.0+%, TCI America™
$123.03 - $415.68
Chemical Identifiers
| CAS | 2836-82-0 |
|---|---|
| Molecular Formula | C9H9FO |
| Molecular Weight (g/mol) | 152.168 |
| MDL Number | MFCD00000325 |
| InChI Key | BANVZEUCJHUPOI-UHFFFAOYSA-N |
| Synonym | 2-fluorophenylacetone, 1-2-fluorophenyl propan-2-one, 1-2-fluorophenyl acetone, 2-fluorophenyl acetone, o-fluorophenylacetone, 2-propanone, 1-2-fluorophenyl, acmc-20anub, ksc497e4p, 1-2-fluorophenyl acetone # |
| PubChem CID | 76086 |
| IUPAC Name | 1-(2-fluorophenyl)propan-2-one |
| SMILES | CC(=O)CC1=CC=CC=C1F |
Chemical Identifiers
| 2836-82-0 | |
| 152.168 | |
| BANVZEUCJHUPOI-UHFFFAOYSA-N | |
| 76086 | |
| CC(=O)CC1=CC=CC=C1F |
| C9H9FO | |
| MFCD00000325 | |
| 2-fluorophenylacetone, 1-2-fluorophenyl propan-2-one, 1-2-fluorophenyl acetone, 2-fluorophenyl acetone, o-fluorophenylacetone, 2-propanone, 1-2-fluorophenyl, acmc-20anub, ksc497e4p, 1-2-fluorophenyl acetone # | |
| 1-(2-fluorophenyl)propan-2-one |
Specifications
| 2836-82-0 | |
| 47°C | |
| MFCD00000325 | |
| 2-fluorophenylacetone, 1-2-fluorophenyl propan-2-one, 1-2-fluorophenyl acetone, 2-fluorophenyl acetone, o-fluorophenylacetone, 2-propanone, 1-2-fluorophenyl, acmc-20anub, ksc497e4p, 1-2-fluorophenyl acetone # | |
| CC(=O)CC1=CC=CC=C1F | |
| 152.168 | |
| 152.17 | |
| Liquid |
| Yellow | |
| C9H9FO | |
| 5 g | |
| BANVZEUCJHUPOI-UHFFFAOYSA-N | |
| 1-(2-fluorophenyl)propan-2-one | |
| 76086 | |
| ≥97.0% (GC) | |
| 2-Fluorophenylacetone |
Safety and Handling
TSCA : No