missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorophenylacetone, 98%

Catalog No. AAH6153506
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6153506 5 g
AAH6153514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6153506 Supplier Thermo Scientific Chemicals Supplier No. H6153506

CAS: 2836-82-0 | C9H9FO | 152.168 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2836-82-0
Molecular Formula C9H9FO
Molecular Weight (g/mol) 152.168
MDL Number MFCD00000325
InChI Key BANVZEUCJHUPOI-UHFFFAOYSA-N
Synonym 2-fluorophenylacetone, 1-2-fluorophenyl propan-2-one, 1-2-fluorophenyl acetone, 2-fluorophenyl acetone, o-fluorophenylacetone, 2-propanone, 1-2-fluorophenyl, acmc-20anub, ksc497e4p, 1-2-fluorophenyl acetone #
PubChem CID 76086
IUPAC Name 1-(2-fluorophenyl)propan-2-one
SMILES CC(=O)CC1=CC=CC=C1F

Specifications

Color Yellow
Density 1.077 g/mL
Boiling Point 47°C
Flash Point 85°C (185°F)
Refractive Index 1.5
Quantity 5 g
Formula Weight 152.17
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Fluorophenylacetone
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 220-627-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only