missing translation for 'onlineSavingsMsg'
Learn More

(2-Fluorophenyl)thiourea 98.0+%, TCI America™

Catalog No. p-7130051
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F10551G 1 g
F10555G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F10551G Supplier TCI America Supplier No. F10551G
Only null left

Chemical Identifiers

CAS 656-32-6
Molecular Formula C7H7FN2S
Molecular Weight (g/mol) 170.205
MDL Number MFCD00041148
InChI Key WYVZQQOFMQRNPF-UHFFFAOYSA-N
Synonym 1-2-fluorophenyl thiourea, 2-fluorophenyl thiourea, 1-2-fluorophenyl-2-thiourea, thiourea, 2-fluorophenyl, n-2-fluorophenyl thiourea, amino 2-fluorophenyl amino methane-1-thione, maybridge1_008774, acmc-1b7vs, 2-fluoro-phenyl-thiourea
PubChem CID 2737500
IUPAC Name (2-fluorophenyl)thiourea
SMILES C1=CC=C(C(=C1)NC(=S)N)F

Specifications

Melting Point 146°C
Color White-Yellow
Quantity 1 g
UN Number 2811
Formula Weight 170.21
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (2-Fluorophenyl)thiourea